ContactUsLinkComponent
Cell Viability and Proliferation Reagent, Biotium
Cell Viability and Proliferation Reagent, Biotium
Catalog # 89138-264
Supplier:  Biotium
CAS Number:  
Cell Viability and Proliferation Reagent, Biotium
Catalog # 89138-264
Supplier:  Biotium
Supplier Number:  10060
CAS Number:  
Specifications
  • Description:
    XTT reagent
  • Size:
    100 mg
  • Cat. No.:
    89138-264
  • Supplier no.:
    10060
Specifications
About this item
XTT reagent is a tetrazolium derivative used widely in cell viability and proliferation testing.
  • Pale yellow solid soluble in water
  • Store desiccated at 4°C
XTT reagent is a tetrazolium derivative used widely in cell viability and proliferation testing. This is readily reduced in viable cells to a highly water-soluble orange colored product. Applications of the reagent include studies of antifungal susceptibility, drug sensitivity of cells, parasitic nematode viability and tumor cell cytotoxicity.