1X PBS (Phosphate Buffered Saline), 0.05% Sodium Azide, Teknova
Catalog # 101446-792
Supplier: Avantor
CAS Number:
Specifications
- Description:1X PBS
- Size:2 L
- Storage Temperature:RT
- Shipping Temperature:Ambient
- Cat. No.:101446-792
- Supplier no.:P0202
Specifications
About this item
1X Phosphate-Buffered Saline with 0.05% Sodium Azide
Suitable for cell culture and molecular biology applications.