Specifications
- Description:Chameleon® Duo Pre-stained Protein Ladder, 500 μL
- Cat. No.:102971-126
- Supplier no.:928-60000
Specifications
About this item
These Pre-stained Protein Ladder provides multi-colored, pre-stained bands for visual inspection and two-color near-infrared detection
- Easy visualization of gel migration and protein size
- Multi-colored, pre-stained bands
- Includes 11 protein bands
Use the ladder for easy visualization of gel migration and protein size, and to orient gel or membrane quickly.
Includes 11 protein bands from 8 to 260 kDa.