Order Entry
United States
ContactUsLinkComponent
Difco™ Bacto M9 Minimal Salts, BD
Difco™ Bacto M9 Minimal Salts, BD
Catalog # 90003-416
Supplier:  BD
CAS Number:  
Difco™ Bacto M9 Minimal Salts, BD
Catalog # 90003-416
Supplier:  BD
Supplier Number:  248510
CAS Number:  

Specifications

  • Description:
    M9 Minimal Salts, Animal Derived Components, pH 7.0 - 7.4 at 25 °C
  • Size:
    500 g
  • Cat. No.:
    90003-416
  • Supplier no.:
    248510

Specifications

About this item

Dehydrated culture media for cultivation of recombinant strains of E.coli.