BD BBL™ Herrold’s Egg Yolk Agar, BD Diagnostics
Supplier: BD
Certificates
About this item
Herrold’s egg yolk agar contains amphotericin B, nalidixic acid and vancomycin
- Selective isolation of Mycobacterium paratuberculosis
- Helps Mycobacterial growth
Agar without mycobactin helps in the growth of Mycobacteria other than Mycobacterium paratuberculosis.
It is used for the selective isolation and differentiation of Mycobacterium paratuberculosis.
Frequently Bought Together