Standard Bore Extention Set, B.Braun
Supplier: B Braun
Certificates
About this item
This standard bore extension set has a female luer connector, and distal SPIN-LOCK connector.
- DEHP- and latex-free
- No injection site