Clarity BG1000 Blood Glucose Meter and Strips, Clarity Diagnostics
Supplier: Clarity Diagnostics
Certificates
About this item
Clarity BG1000 blood glucose meter cleared by the FDA for single and multi-patient use.
- High reliability
- Can be used with capillary whole blood samples
- Pre-meal/post meal and control marker
- Sophisticated software for patient management
- Internal memory of 300 results
- No code meter
Unique strip ejector at the back of the meter.
Certifications: AST approved. ISO15197 compliant.
Accessories information: Clarity BG1000 blood glucose meter strips are for use with with the Clarity BG1000 Meter. Requires a small sample size of just 0.5µl whole blood. Test time of 5 seconds. AST approved for fingertip, palm, forearm. Measuring range is 20-600 mg/dl. HCT range is 20-60%. Based on electrochemical biosensor technology.
Clarity BG1000 glucose control solutions are for use with the Clarity BG1000 Meter. 90 day open vial stability. 1 vial of normal and 1 vial of high control. 4 ml of control of each level. Room temperature storage.
Clarity BG1000 glucose control solutions are for use with the Clarity BG1000 Meter. 90 day open vial stability. 1 vial of normal and 1 vial of high control. 4 ml of control of each level. Room temperature storage.
Frequently Bought Together