
Sulfuric acid 0.25 M H2SO4 (0.5N) in aqueous solution ≥99.9% Reag. Ph. Eur. volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08
Boiling Pt:>100 °C (1.013 hPa)
Density:1.020 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-755-4
UN:3264
ADR:8,III
:GHS05
Specification Test Results
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
Related Information
Frequently Bought Together