Order Entry
United States
ContactUsLinkComponent
Sulfuric acid 1 M H2SO4 (2N) volumetric solution, Fluka™
Corrosive
Sulfuric acid 1 M H2SO4 (2N) volumetric solution, Fluka™
Catalog # BJ35276-1L
CAS Number:  
Corrosive
Sulfuric acid 1 M H2SO4 (2N) volumetric solution, Fluka™
Catalog # BJ35276-1L
Supplier Number:  35276-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 L
  • Cat. No.:
    BJ35276-1L
  • Supplier no.:
    35276-1L

Specifications

Specification Test Results

  • Factor:
    0.999 - 1.001
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :98.08
Density:1.05 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3264
ADR:8,III
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589