Sodium hydroxide 0.5 M NaOH (0.5N) volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:>100 °C (1.013 hPa)
Melting Pt:319 °C
Density:1.020 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1824
ADR:8,III
:GHS05
Specification Test Results
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
Related Information
Frequently Bought Together