Potassium permanganate 0.02 M KMnO4 (0.1N) volumetric solution, Fluka™
Catalog # BJ35186-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:BJ35186-1L
- Supplier no.:35186-1L
Specifications
Specification Test Results
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
About this item
:
:158.03 g/mol
Storage Temperature:Ambient
UN:3082
ADR:9,III
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364