Order Entry
United States
ContactUsLinkComponent
Potassium hydroxide 0.5 M KOH (0.5N) volumetric solution, Fluka™
Corrosive
Potassium hydroxide 0.5 M KOH (0.5N) volumetric solution, Fluka™
Catalog # BJ35116-1L
CAS Number:  
Corrosive
Potassium hydroxide 0.5 M KOH (0.5N) volumetric solution, Fluka™
Catalog # BJ35116-1L
Supplier Number:  35116-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 L
  • Cat. No.:
    BJ35116-1L
  • Supplier no.:
    35116-1L

Specifications

Specification Test Results

  • Factor:
    0.999 - 1.001
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :56.11 g/mol
Boiling Pt: 100 °C
Density:1.023 g/cm³
Storage Temperature:Ambient
UN:1814
ADR:8,II
 :GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553