
Potassium hydroxide 1 M KOH (1N) volumetric solution, Fluka™
Catalog # BJ35113-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 L
- Cat. No.:BJ35113-1L
- Supplier no.:35113-1L
Specifications
Specification Test Results
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
About this item
:
:56.11 g/mol
Boiling Pt: 100 °C
Density:1.05 g/cm³
Storage Temperature:Ambient
UN:1814
ADR:8,II
:GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553