Order Entry
United States
ContactUsLinkComponent
Iodine 0.5 M I2 (1.0N) in aqueous solution volumetric solution, Fluka™
Irritant
Health
Iodine 0.5 M I2 (1.0N) in aqueous solution volumetric solution, Fluka™
Catalog # BJ-35089-1L
CAS Number:  
Irritant
Health
Iodine 0.5 M I2 (1.0N) in aqueous solution volumetric solution, Fluka™
Catalog # BJ-35089-1L
Supplier Number:  35089-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    BJ-35089-1L

Specifications

Specification Test Results

  • Factor:
    0.999 - 1.001

Specification Test Results

About this item

 :
 :253.81 g/mol
Boiling Pt:>100 °C (1.013 hPa)
Melting Pt:113,5 °C
Density:1.260 g/cm³ (20 °C)
UN:3495
ADR:8,III
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355