Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Magnesium sulphate
Magnesium sulfate 0.1 M volumetric solution, Fluka™
Magnesium sulfate 0.1 M volumetric solution, Fluka™
Magnesium sulfate 0.1 M volumetric solution, Fluka™
BJ34276-1L
:
Honeywell Research Chemicals
:
Magnesium sulfate 0.1 M volumetric solution, Fluka™
BJ34276-1L
:
Honeywell Research Chemicals
:
34276-1L
:
Pack type:
Plastic bottle
Size:
1 L
Cat. No.:
BJ34276-1L
Supplier no.:
34276-1L
Factor:
0.998 - 1.002
:
:
120.37 g/mol
:
Ambient
:
7487-88-9
:
MgSO₄
:
MFCD00011110