VeriSpec™ Nexion AFT ICP-MS Detection Solution, 13 Components, RICCA Chemical Company
Catalog # RCRV010889100N
Supplier: Ricca Chemical
CAS Number:
Specifications
- Pack type:Plastic Bottle
- Size:100 ml
- Cat. No.:RCRV010889100N
- Supplier no.:RV010889-100N
Specifications
About this item
Certified reference material (CRM) for ICP-MS
50 ppb: Al, Ba, Ce, Co, In, Li, Mg, Mn, Ni, Pb, Tb, U, Zn
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility