Order Entry
United States
ContactUsLinkComponent
VeriSpec® Tungsten Standard for ICP/MS 10 ppm NH₄OH, RICCA Chemical Company
VeriSpec® Tungsten Standard for ICP/MS 10 ppm NH₄OH, RICCA Chemical Company
Catalog # RCRV010634100N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec® Tungsten Standard for ICP/MS 10 ppm NH₄OH, RICCA Chemical Company
Catalog # RCRV010634100N
Supplier:  Ricca Chemical
Supplier Number:  RV010634-100N
CAS Number:  

Specifications

  • Concentration:
    10 µg/ml
  • Matrix:
    H₂O/tr. NH₄OH
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. No.:
    RCRV010634100N
  • Element:
    Tungsten
  • Supplier no.:
    RV010634-100N

Specifications

About this item

Certified reference material (CRM) for ICP-MS

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility