VeriSpec™ Praseodymium Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
Catalog # RCRV010543100N
Supplier: Ricca Chemical
CAS Number:
Specifications
- Concentration:100 µg/ml
- Matrix:2% HNO₃
- Pack type:Plastic Bottle
- Size:100 ml
- Cat. No.:RCRV010543100N
- Element:Praseodymium
- Supplier no.:RV010543-100N
Specifications
About this item
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility