VeriSpec® Selenium (Se) Standard for AAS 20 ppb in 1% HNO₃, RICCA Chemical Company
Catalog # RCRV010253-50N
Supplier: Avantor
CAS Number:
Specifications
- Concentration:20 ppb
- Matrix:1% HNO₃
- Pack type:Plastic Bottle
- Size:50 ml
- Cat. No.:RCRV010253-50N
- Element:Selenium
- Supplier no.:RV010253-50N
Specifications
About this item
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility