Order Entry
United States
ContactUsLinkComponent
VeriSpec® Cadmium (Cd) Standard for AAS 2 ppb in 1% HNO₃, RICCA Chemical Company
VeriSpec® Cadmium (Cd) Standard for AAS 2 ppb in 1% HNO₃, RICCA Chemical Company
Catalog # RCRV010123-50N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec® Cadmium (Cd) Standard for AAS 2 ppb in 1% HNO₃, RICCA Chemical Company
Catalog # RCRV010123-50N
Supplier:  Ricca Chemical
Supplier Number:  RV010123-50N
CAS Number:  

Specifications

  • Concentration:
    2 ppb
  • Matrix:
    1% HNO₃
  • Pack type:
    Plastic Bottle
  • Size:
    50 ml
  • Cat. No.:
    RCRV010123-50N
  • Element:
    Cadmium
  • Supplier no.:
    RV010123-50N

Specifications

About this item

Certified reference material (CRM) for AA (atomic absorption)

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility