ContactUsLinkComponent
Western Blot Strip-it™ Buffer, Advansta
Western Blot Strip-it™ Buffer, Advansta
Catalog # 103254-874
Supplier:  ADVANSTA INC MS
CAS Number:  
Western Blot Strip-it™ Buffer, Advansta
Catalog # 103254-874
Supplier:  ADVANSTA INC MS
Supplier Number:  R-03722-D50
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Western Blot Strip-it™ Buffer
  • Size:
    1x 500 mL
  • Cat. No.:
    103254-874
  • Supplier no.:
    R-03722-D50
Specifications
About this item
Stripping buffer for stripping antibodies from chemiluminescent Western blots.
  • Conserve valuable samples by probing the same blot multiple times
  • Quickly strip and prepare for reprobing in 25 minutes
  • The gentle protocol removes antibodies without harsh reagents or elevated temperatures
  • Save time by re-probing the same blot
  • Compatible with both nitrocellulose and PVDF membranes and with a variety of chemiluminescent detection reagents
Advansta's Western Blot Strip-It Buffer uniformly removes antibodies from developed Western blots so the blot can be reprobed with another set of antibodies. Stripping and reprobing saves precious samples and the entire stripping protocol takes only 25 minutes. The gentle, odor-free buffer removes bound antibodies without harsh reagents or elevated temperatures and will not damage antigens or membranes.
Conserve your precious sample by reprobing one blot multiple times instead of running duplicate blots. Advansta's Western Blot Strip-It Buffer provides uniform stripping so you can obtain high-quality, qualitative data with multiple probes of the same blot. Strip-It is also compatible with most chemiluminescent reagents.