Orders LinkContactUsLinkComponent
Protein Sample Loading Buffers, Advansta
Protein Sample Loading Buffers, Advansta
Marketsource
Supplier:   ADVANSTA INC MS
Protein Sample Loading Buffers, Advansta
Supplier:  ADVANSTA INC MS
Marketsource
Certificates
Protein Sample Loading Buffers, Advansta
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pre-mixed standard Laemmli loading buffers for protein electrophoresis.
  • Pre-mixed - just add equal volume to your protein sample
  • Obtain reproducible results with standardized buffers
  • Choose from reducing or non-reducing loading buffer
These ready-to-use Laemmli loading buffers contain Bromophenol blue to enable visualization of the dye front and monitoring of the progress of electrophoresis.
Choose from reducing or non-reducing buffers, each supplied as a 2X concentrate.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 103254-792
    Non-reducing protein sample loading buffer (2X)
    Non-reducing protein sample loading buffer (2X)
    1x 1 mL
    Log in for availability
    Log in for availability
    Each (1ml)
    Each (1ml) - $23.08
    $23.08
    Each (1ml)
       $23.08
    Supplier Number: R-03018-B10
    Specifications:
    Marketsource
    Marketsource
  • 103254-794
    Non-reducing protein sample loading buffer (2X)
    Non-reducing protein sample loading buffer (2X)
    1x 5 mL
    Log in for availability
    Log in for availability
    Each (5ml)
    Each (5ml) - $73.85
    $73.85
    Each (5ml)
       $73.85
    Supplier Number: R-03018-B50
    Specifications:
    Marketsource
    Marketsource
  • 103254-914
    Reducing protein sample loading buffer (2X)
    Reducing protein sample loading buffer (2X)
    1x 1 mL
    Log in for availability
    Log in for availability
    Each (1ml)
    Each (1ml) - $31.15
    $31.15
    Each (1ml)
       $31.15
    Supplier Number: R-03019-B10
    Specifications:
    Marketsource
    Marketsource
  • 103254-916
    Reducing protein sample loading buffer (2X)
    Reducing protein sample loading buffer (2X)
    1x 5 mL
    Log in for availability
    Log in for availability
    Each (5ml)
    Each (5ml) - $102.69
    $102.69
    Each (5ml)
       $102.69
    Supplier Number: R-03019-B50
    Specifications:
    Marketsource
    Marketsource