SP Wilmad-LabGlass Hemispherical Micro Heating Mantles, SP Industries
Supplier: SP Industries
Certificates
About this item
These micro heating mantles are made of soft, all glass fabric and support the bottom half of glass flasks
Mantles allow for full view of the contents of single or multi-neck flasks. Include a separable 3' long, 2-wire cord and locking connector.
Frequently Bought Together