To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Secondary
- Antigen Name:Ig
- Antigen Symbol:Ig
- Clonality:Monoclonal
- Clone:SB87a
- Conjugation:Spectral Red® RPE (R-Phycoerythrin)/Cy5®
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Rabbit
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:0.5 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Rabbit IgG and IgM
- Cat. No.:101442-850
- Supplier no.:4065-13
Specifications
About this item
PE/CY5 (SPRD) Anti-Rabbit Ig antibody for use in flow cytometry assays.
Type: Secondary
Antigen: Ig
Clonality: Monoclonal
Clone: SB87a
Conjugation: Spectral Red® RPE (R-Phycoerythrin)/Cy5®
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Rabbit