Order Entry
United States
ContactUsLinkComponent
Anti-TNF-alpha Rat Monoclonal Antibody (Biotin) [clone: MP6-XT3]
Anti-TNF-alpha Rat Monoclonal Antibody (Biotin) [clone: MP6-XT3]
Catalog # 101442-620
Anti-TNF-alpha Rat Monoclonal Antibody (Biotin) [clone: MP6-XT3]
Catalog # 101442-620
Supplier Number:  10229-08
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    TNF-alpha
  • Clonality:
    Monoclonal
  • Clone:
    MP6-XT3
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.5 mg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    21926
  • Antigen Synonyms:
    TNFSF-2|DIF|macrophage cytotoxic factor |cachectin|differentiation inducing factor|necrosin|MCF|Tumor necrosis factor-α
  • Storage Buffer:
    Supplied in PBS/NaN3
  • Storage Temperature:
    2-8°C
  • Concentration:
    0.5 mg/mL
  • Shipping Temperature:
    Ice Packs
  • Immunogen:
    E. coli-expressed mouse TNF-α
  • Tested Applications:
    ELISPOT, Multiplex
  • Cat. No.:
    101442-620
  • Supplier no.:
    10229-08

Specifications

About this item

Biotin Anti-Mouse TNF-alpha antibody for use in flow cytometry, immunohistochemistry, western blotting, ELISA, ELISpot, and multiplex assays.

Type: Primary
Antigen: TNF-alpha
Clonality: Monoclonal
Clone: MP6-XT3
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse