To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin M
- Antigen Symbol:IgM
- Clonality:Monoclonal
- Clone:SA-DA4
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:0.5 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:3507
- Storage Buffer:Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
- Storage Temperature:2-8°C
- Concentration:0.5 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Human IgM myeloma protein
- Tested Applications:Multiplex, Stim
- Cat. No.:100245-638
- Supplier no.:9020-01
Specifications
About this item
Purified Anti-Human IgM antibody for use in ELISA, flow cytometry, immunohistochemistry / immunocytochemistry, western blotting, immunoprecipitation, stimulation, and multiplex assays.
Type: Secondary
Antigen: IgM
Clonality: Monoclonal
Clone: SA-DA4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human