Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:CD44
- Antigen Symbol:CD44
- Clonality:Monoclonal
- Clone:AV6
- Conjugation:Spectral Red® RPE (R-Phycoerythrin)/Cy5®
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Turkey,Chicken,Pigeon
- Size:0.1 mg
- Form:Liquid
- Gene ID:395666
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:0.1 mg/mL
- Shipping Temperature:Ambient Temperature
- Cat. No.:100245-614
- Supplier no.:8400-13
Chicken CD44 is a type I transmembrane glycoprotein expressed on many cell types including B cells, most T cells, monocytes, and some epithelial cells. CD44 primarily functions as an adhesion molecule. It has been implicated in various processes such as hematopoiesis, lymphocyte homing, leukocyte activation, tumor metastasis, and development. The monoclonal antibody AV6 also reacts with turkey peripheral blood mononuclear cells.
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: AV6
Conjugation: Spectral Red® RPE (R-Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Turkey, Pigeon