To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD4
- Antigen Symbol:CD4
- Clonality:Monoclonal
- Clone:74-12-4
- Conjugation:RPE (R-Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Pig
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:404704
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:0.1 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Fresh dd miniature swine thymocytes
- Cat. No.:100244-848
- Supplier no.:4515-09
Specifications
About this item
Porcine CD4 is a type I transmembrane glycoprotein and a member of the immunoglobulin superfamily of cell surface receptors. CD4 is an accessory molecule in the recognition of foreign antigens in association with MHC Class II molecules by T lymphocytes.
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 74-12-4
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Porcine