About this item
The 3C4 monoclonal antibody reacts with the cell surface glycoprotein mouse CD102 (ICAM-2), a ligand for the leucocyte integrin CD11a/CD18 (LFA-1). CD102 is constitutively expressed on T cells, B cells, and at high levels on vascular endothelial cells. It is also expressed on a variety of leukocyte cell lines. The 3C4 antibody blocks the interaction between ICAM-2 and LFA-1.
Type: Primary
Antigen: CD102
Clonality: Monoclonal
Clone: 3C4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 100244-174
- 100244-176
- Antigen Symbol:
- CD102
- CD102
- Antigen Name:
- CD102
- CD102
- Antigen Synonyms:
- ICAM-2|intercellular adhesion molecule-2
- ICAM-2|intercellular adhesion molecule-2
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- 3C4
- 3C4
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 15896
- 15896
- Isotype:
- IgG2a
- IgG2a
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- COS cells transfected with mouse ICAM-2 cDNA and BW5147 AKR/J mouse thymoma cell line
- COS cells transfected with mouse ICAM-2 cDNA and BW5147 AKR/J mouse thymoma cell line
- Form:
- Liquid
- Liquid
- Concentration:
- 0.5 mg/mL
- 0.1 mg/mL
- Storage Buffer:
- Supplied in PBS/NaN3
- Supplied in PBS/NaN3
- Storage Temperature:
- 2-8°C
- 2-8°C
- Shipping Temperature:
- Ambient Temperature
- Ambient Temperature