To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:CD49b
- Antigen Symbol:CD49b
- Clonality:Monoclonal
- Clone:DX5
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM
- Reactivity:Mouse
- Size:0.5 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:16398
- Antigen Synonyms:very late antigen-2|α2 integrin|VLA-2α|pan NK
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:0.5 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:NK cells isolated from C57BL/6 mice
- Cat. No.:100243-874
- Supplier no.:1806-02
The monoclonal antibody DX5 reacts with CD49b, also known as very late antigen-2 (VLA-2) and α2 integrin. The antibody stains the majority of NK cells and a small subpopulation of T cells in all mouse strains tested (e.g., A/J, AKR, BALB/c, C3H/HeJ, C57BL/6, C57BL/10, C57BR, C58, CBA/Ca, CBA/J, DBA/1, DBA/2, SJL, SWR). Three-color flow cytometry analysis has revealed that most cells that express NK-1.1 (NKR-P1C) also express CD49b. However, small subsets of DX5+NK-1.1- and DX5-NK-1.1+ cells can be found, especially among the CD3+ population of T lymphocytes. NK cells cultured in the presence of IL-2 progressively lose reactivity with DX5 as a consequence of cellular proliferation. Therefore, DX5 can be used to define functionally distinct subsets of murine NK cells. The monoclonal antibody DX5 has not been demonstrated to have activating or blocking activity.
Type: Primary
Antigen: CD49b
Clonality: Monoclonal
Clone: DX5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Mouse