To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD14 molecule
- Antigen Symbol:CD14
- Clonality:Monoclonal
- Clone:UCHM-1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Cynomolgus monkey,Human
- Size:100 tests
- Environmentally Preferable:
- Form:Liquid
- Gene ID:929
- Antigen Synonyms:gp55|LPS-R|LPS receptor
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:Lot specific
- Shipping Temperature:Ambient Temperature
- Immunogen:Thymocytes and peripheral blood lymphocytes from a remission AML patient
- Cat. No.:100246-156
- Supplier no.:9560-02
Specifications
About this item
CD14 is a 55 kDa glycosphosphatidylinositol (GPI)-anchored single chain glycoprotein. It is expressed on monocytes, macrophages, dendritic cells, and Langerhans cells. It is also weakly expressed on neutrophils. CD14 appears to be involved in clearance of gram-negative bacteria via its high affinity binding to LPS-LPB complexes.
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: UCHM-1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Cynomolgus