To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD19 molecule
- Antigen Symbol:CD19
- Clonality:Monoclonal
- Clone:SJ25-C1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 tests
- Environmentally Preferable:
- Form:Liquid
- Gene ID:930
- Antigen Synonyms:Leu-12|B-lymphocyte antigen|B4
- Antibody Fragment:F(ab')2
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:Lot specific
- Shipping Temperature:Ambient Temperature
- Immunogen:NALM-1 and NALM-16 leukemia cell line
- Cat. No.:100245-912
- Supplier no.:9342-02
Specifications
About this item
CD19 is a type I transmembrane glycoprotein and a member of the immunoglobulin superfamily. It is expressed at all stages of B cell differentiation except terminally differentiated plasma cells. It is also present on the cell surface of follicular dendritic cells. CD19 associates with CD21, CD81, Leu 13 and/or MHC Class II molecules to form a signal transduction complex on the B cell surface. This signaling complex modulates the activation threshold for the B cell antigen receptor (BCR).
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25-C1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human