To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:B Cell Subset
- Antigen Symbol:B Cell Subset
- Clonality:Monoclonal
- Clone:BB6-10A10
- Conjugation:RPE (R-Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Pig
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:0.1 mg/mL
- Shipping Temperature:Ambient Temperature
- Cat. No.:100244-884
- Supplier no.:4550-09
The monoclonal antibody BB6-10A10 stains the following percentages of surface immunoglobulin (sIg) positive cells in porcine lymphoid tissues - 11-15% in peripheral blood, 22-23% in mesenteric lymph nodes, 79-87% in ileal Peyer’s patches (ILPP), 6-17 in spleen, and 0% in thymus. While identification of the antigen(s) that are found predominantly on ILPP B cells and which are recognized by BB6-10A10 is speculative, the antibody appears to react with an immature population of B cells in the follicles of ILPPs and with a subpopulation of sIglow B cells in lymph nodes.
Type: Primary
Antigen: B Cell Subset
Clonality: Monoclonal
Clone: BB6-10A10
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Porcine