Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:CD3 epsilon
- Antigen Symbol:CD3epsilon
- Clonality:Monoclonal
- Clone:BB23-8E6
- Conjugation:Biotin
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Pig
- Size:0.5 mg
- Form:Liquid
- Gene ID:397455
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:0.5 mg/mL
- Shipping Temperature:Ambient Temperature
- Cat. No.:100244-838
- Supplier no.:4511-08
Specifications
About this item
Porcine CD3ε is a member of the T-cell receptor-associated CD3 complex. It is found on a subpopulation of thymocytes and on all pig T lymphocytes.
Type: Primary
Antigen: CD3epsilon
Clonality: Monoclonal
Clone: BB23-8E6
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Porcine