Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:CD1
- Antigen Symbol:CD1
- Clonality:Monoclonal
- Clone:76-7-4
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Pig
- Size:0.5 mg
- Form:Liquid
- Gene ID:396800
- Antigen Synonyms:CD1.1|CD1c
- Storage Buffer:Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
- Storage Temperature:2-8°C
- Concentration:0.5 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Fresh dd miniature swine thymocytes
- Tested Applications:CMCD
- Cat. No.:100244-816
- Supplier no.:4500-01
Specifications
About this item
Porcine CD1 is a type I transmembrane glycoprotein and a member of the immunoglobulin superfamily of cell surface receptors. It has a domain organization similar to that of MHC class I molecules and is expressed in association with β2-microglobulin. CD1 is found on B cells, macrophages, and immature thymocytes. There is evidence for a role of CD1 in presentation of lipids and peptides to T cells.
Type: Primary
Antigen: CD1
Clonality: Monoclonal
Clone: 76-7-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Porcine