Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:H-2Kk
- Antigen Symbol:H-2Kk
- Clonality:Monoclonal
- Clone:16-3-22S
- Conjugation:Biotin
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Size:0.5 mg
- Form:Liquid
- Gene ID:14972
- Antigen Synonyms:MHC Class I
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:0.5 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:C3H mouse splenocytes
- Cat. No.:100244-148
- Supplier no.:1916-08
The monoclonal antibody 16-3-22S reacts with the H-2Kk MHC class I alloantigen. Cross-reactivity with splenocytes of SJL/Hsd mice has been observed by flow cytometry. The antibody does not react with other (e.g., b, d, q) haplotypes.
Type: Primary
Antigen: H-2Kk
Clonality: Monoclonal
Clone: 16-3-22S
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse