To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD94
- Antigen Symbol:CD94
- Clonality:Monoclonal
- Clone:18d3
- Conjugation:RPE (R-Phycoerythrin)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:16643
- Antigen Synonyms:Kp43|KLRD1
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:0.1 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:CHO cells transfected with the B6 allele of CD94
- Cat. No.:100243-904
- Supplier no.:1809-09
Specifications
About this item
The monoclonal antibody 18d3 reacts with the mouse CD94 receptor subunit, a type II transmembrane glycoprotein. CD94 is expressed on the surface of NK cells, NK T cells, and some memory phenotype CD8+ cells. When CD94 is paired with NKG2A, C, or E subunit, it binds the nonclassical MHC class Ib molecule Qa-1b and results in a CD94high population by flow cytometry. Approximately half of NK cells do not express NKG2A, C or E; these cells express CD94 on the surface in a form that does not bind Qa-1b and is believed to be nonfunctional, producing a CD94low population by flow cytometry. CD94 is thought to play a role in the adhesion and activation of the NK cell lineage.
Type: Primary
Antigen: CD94
Clonality: Monoclonal
Clone: 18d3
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse