To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Ly-49C/F/H/I
- Antigen Symbol:Ly-49C/F/H/I
- Clonality:Monoclonal
- Clone:14B11
- Conjugation:RPE (R-Phycoerythrin)
- Flow Cytometry:Yes
- Host:Hamster
- Isotype:IgG
- Reactivity:Mouse
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:16634/16637/16639/16640
- Antigen Synonyms:Ly-49
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:0.1 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:IL-2 activated natural killed (A-LAK) cells from C57BL/6 mice
- Cat. No.:100243-830
- Supplier no.:1801-09
Specifications
About this item
The monoclonal antibody 14B11 reacts with murine Ly49C, Ly49F, Ly49H, and Ly49I, members of the lectin-like homodimeric Ly49 family of cell surface receptors. Mouse Ly49 receptors exhibit allelic specificity for MHC class I Ia molecules and are thought to serve to prevent natural killer (NK) cells from attacking normal cells while allowing them to attack infected or transformed cells in which class I molecules have been downregulated. These inhibitory receptors are also expressed on a subpopulation of mouse CD8+ T cells. The 14B11 antibody stains ~90% of NK1.1+CD3- splenocytes and ~6% of CD8+ spleen cells from C57BL/6 mice. The antibody also stains 30-80% of NK1.1+CD3- splenocytes in other mouse strains tested (BALB/c, C3H, 129, SJL, AKR/J).
Type: Primary
Antigen: Ly-49C/F/H/I
Clonality: Monoclonal
Clone: 14B11
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse