Order Entry
United States
ContactUsLinkComponent
Anti-TCRgammadelta Hamster Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: UC7-13D5]
Anti-TCRgammadelta Hamster Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: UC7-13D5]
Certificates
Anti-TCRgammadelta Hamster Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: UC7-13D5]

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Catalog No:
  • 100243-752
  • 100243-750
  • Antigen Symbol:
  • TCR gamma delta
  • TCR gamma delta
  • Antigen Name:
  • TCR gamma delta
  • TCR gamma delta
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • RPE (R-Phycoerythrin)
  • RPE (R-Phycoerythrin)
  • Clone:
  • UC7-13D5
  • UC7-13D5
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Hamster
  • Hamster
  • Gene ID:
  • 110066/110067
  • 110066/110067
  • Isotype:
  • IgG
  • IgG
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Mouse T cell clone G8
  • Mouse T cell clone G8
  • Form:
  • Liquid
  • Liquid
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • Supplied in PBS/NaN3 and a stabilizing agent
  • Supplied in PBS/NaN3 and a stabilizing agent
  • Storage Temperature:
  • 2-8°C
  • 2-8°C
  • Shipping Temperature:
  • Ambient Temperature
  • Ambient Temperature

Specifications

Frequently Bought Together