Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:CD69
- Antigen Symbol:CD69
- Clonality:Monoclonal
- Clone:H1.2F3
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Hamster
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Size:0.5 mg
- Form:Liquid
- Gene ID:12515
- Antigen Synonyms:EA1|MLR3|gp34/28|AIM|VEA|very early activation antigen|Leu-23
- Storage Buffer:Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
- Storage Temperature:2-8°C
- Concentration:0.5 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Mouse dendritic epidermal cell line Y245
- Tested Applications:Costim, Activ
- Cat. No.:100243-606
- Supplier no.:1715-01
Specifications
About this item
CD69, also known as very early activation (VEA) antigen, is a disulfide-linked transmembrane homodimer whose differentially glycosylated subunits range from 35-39 kDa. It is a C-type lectin most closely related to the NKR-P1 and Ly-49 NK cell-activation molecules. CD69 is widely expressed on hematopoietic cells, including lymphocytes, neutrophils and eosinophils. Although not detectable on resting lymphocytes, its expression is rapidly upregulated upon activation of T, B and NK cells, and neutrophils. Constitutive expression of CD69 on subsets of thymocytes suggests that it may be involved in regulation of developmental events in addition to its role in activation of a variety of hematopoietic cells. The monoclonal antibody H1.2F3 augments PMA-induced T-cell proliferation and induces redirected lysis of Fc receptor-bearing target cells by NK cells.
Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: H1.2F3
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse