Order Entry
United States
ContactUsLinkComponent
Anti-CD11b Rat Monoclonal Antibody (RPE (R-Phycoerythrin)/Cy7®) [clone: 3A33]
Anti-CD11b Rat Monoclonal Antibody (RPE (R-Phycoerythrin)/Cy7®) [clone: 3A33]
Catalog # 100243-224
Anti-CD11b Rat Monoclonal Antibody (RPE (R-Phycoerythrin)/Cy7®) [clone: 3A33]
Catalog # 100243-224
Supplier Number:  1560-17

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    CD11b
  • Antigen Symbol:
    CD11b
  • Clonality:
    Monoclonal
  • Clone:
    3A33
  • Conjugation:
    RPE (R-Phycoerythrin)/Cy7®
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Size:
    0.1 mg
  • Form:
    Liquid
  • Gene ID:
    16409
  • Antigen Synonyms:
    C3biR|Mo1|Integrin αM|CR3|Mac-1|Mac-1α
  • Storage Buffer:
    Supplied in PBS/NaN3 and a stabilizing agent
  • Storage Temperature:
    2-8°C
  • Concentration:
    0.1 mg/mL
  • Shipping Temperature:
    Ambient Temperature
  • Immunogen:
    Peritoneal macrophages from B6D2 hybrid mice
  • Cat. No.:
    100243-224
  • Supplier no.:
    1560-17

Specifications

About this item

CD11b (integrin αM subunit) combines with CD18 (integrin β2 subunit) to form the integrin Mac-1, also known as complement receptor 3 (CR3), which mediates adhesion to C3bi and ICAM-1 (CD54). Mac-1 is expressed at varying levels on granulocytes, macrophages, dendritic cells, NK cells, and B-1 cells in the peritoneal and pleural cavities. In addition to its role in binding C3bi on opsonized targets and mediation of the subsequent ingestion process, Mac-1 is important as an adhesion molecule in the transendothelial migration of monocytes and neutrophils.

Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: 3A33
Conjugation: RPE (R-Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse