About this item
CD11b (integrin αM subunit) combines with CD18 (integrin β2 subunit) to form the integrin Mac-1, also known as complement receptor 3 (CR3), which mediates adhesion to C3bi and ICAM-1 (CD54). Mac-1 is expressed at varying levels on granulocytes, macrophages, dendritic cells, NK cells, and B-1 cells in the peritoneal and pleural cavities. In addition to its role in binding C3bi on opsonized targets and mediation of the subsequent ingestion process, Mac-1 is important as an adhesion molecule in the transendothelial migration of monocytes and neutrophils.
Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: 3A33
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 100243-208
- 100243-206
- Antigen Symbol:
- CD11b
- CD11b
- Antigen Name:
- CD11b
- CD11b
- Antigen Synonyms:
- C3biR|Mo1|Integrin αM|CR3|Mac-1|Mac-1α
- C3biR|Mo1|Integrin αM|CR3|Mac-1|Mac-1α
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- 3A33
- 3A33
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 16409
- 16409
- Isotype:
- IgG2a
- IgG2a
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Peritoneal macrophages from B6D2 hybrid mice
- Peritoneal macrophages from B6D2 hybrid mice
- Form:
- Liquid
- Liquid
- Concentration:
- 0.1 mg/mL
- 0.5 mg/mL
- Storage Buffer:
- Supplied in PBS/NaN3
- Supplied in PBS/NaN3
- Storage Temperature:
- 2-8°C
- 2-8°C
- Shipping Temperature:
- Ambient Temperature
- Ambient Temperature