Nocardia Hydrolysis Kit for Actinomycetes, Hardy Diagnostics
Catalog # 75860-340
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:Nocardia Hydrolysis Kit, HDx
- Cat. No.:75860-340
- Tube size:20×125 mm
- Supplier no.:Z604
Specifications
About this item
For use in determining the hydrolysis reaction on casein, xanthine, hypoxanthine, and tyrosine by Nocardia, Actinomycetales, Actinomadura, and Streptomyces spp.
- Determine the hydrolysis reaction
Packaging: Total of 20 tubes of four different media types (Five each of Casein, Xanthine, Hypoxanthine, and Tyrosine Agars).
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.