Blotting Sponge Pads, Advansta
About this item
Transfer sponge pads for more reproducible Western blot transfers.
- Allows proper pressure to be maintained in transfer sandwiches, resisting compression
- Retains thickness to reuse over multiple experiments
- Conveniently available in two sizes for different transfer apparatuses
- Standardized with uniform flat construction to ensure reproducible and consistent transfers
Advansta's Blotting sponge pads maintain secure contact between gel and membrane in transfer sandwiches and cassettes for consistent transfers. The blotting sponge pads resist compression and are available in two sizes (8 x 11 cm and 9 x 15 cm) to work with multiple transfer setups and membrane sizes.
2 Options Available Below
Product Details & Documents
Transfer sponge pads for more reproducible Western blot transfers.
- Allows proper pressure to be maintained in transfer sandwiches, resisting compression
- Retains thickness to reuse over multiple experiments
- Conveniently available in two sizes for different transfer apparatuses
- Standardized with uniform flat construction to ensure reproducible and consistent transfers
Advansta's Blotting sponge pads maintain secure contact between gel and membrane in transfer sandwiches and cassettes for consistent transfers. The blotting sponge pads resist compression and are available in two sizes (8 x 11 cm and 9 x 15 cm) to work with multiple transfer setups and membrane sizes.
Product Family Options
Product Information
- DimensionsAvailabilityPrice
- 103254-870Blotting Sponge PadsSupplier Number: L-07129-004Specifications:MarketsourceMarketsource
- 103254-872Blotting Sponge PadsSupplier Number: L-07130-004Specifications:MarketsourceMarketsource