Orders LinkContactUsLinkComponent
Development Folders, Advansta
Development Folders, Advansta
Marketsource
Supplier:   ADVANSTA INC MS
Development Folders, Advansta
Supplier:  ADVANSTA INC MS
Marketsource
Certificates
Development Folders, Advansta
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Transparent plastic supports for handling and imaging blots.
  • Get clear images by avoiding wrinkling that can happen with plastic wrap
  • Suitable for all blots, both chemiluminescence and fluorescence imaging
  • Allows safe blot handling, avoiding scratching or ripping
  • Conveniently sized to image up to 2 blots in one folder
Advansta's Blot Development Folders are transparent plastic folders that hold your blot while it is exposed to film or a CCD camera. These folders keep your blot flat for perfect film exposures, avoiding the wrinkling or bunching that can occur with plastic wrap.
The folders are not autofluorescent and are compatible with chemiluminescence and fluorescence imaging. The folders also protect blots during handling.
Product Family Options
Product Information
  • Dimensions
    Size
    Availability
    Price
  • 103254-866
    Blot Development Folders
    Blot Development Folders
    5 x 7"
    25 folders
    Log in for availability
    Log in for availability
    Each
    Each - $69.23
    $69.23
    Each
       $69.23
    Supplier Number: L-07020-025
    Specifications:
    Marketsource
    Marketsource
  • 103254-868
    Blot Development Folders
    Blot Development Folders
    5 x 7"
    100 folders
    Log in for availability
    Log in for availability
    Each
    Each - $132.69
    $132.69
    Each
       $132.69
    Supplier Number: L-07020-100
    Specifications:
    Marketsource
    Marketsource