Orion™ Epoxy ATC Probe, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Get fast, accurate measurements in the laboratory or field.
The Orion™ Epoxy ATC Probe automatically corrects pH measurements for variation in electrode slope due to temperature change.
Ordering information: For temperature readings only. pH probe is sold separately.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.