RapidClean, Advansta
About this item
Rapid clean is the fast and easy way to remove protein from RNA and DNA samples.
- Complete removal of proteins from aqueous solutions of nucleic acids
- Protocol takes less than 5 min
- Provides non-toxic alternative to phenol-chloroform extraction
- Nucleic acid recovery rates >95%
- Recovers nucleic acids from 5 to greater than 40 kb
- Suitable for DNA, RNA, cDNA, microRNA, and oligonucleotides
In less than 5 minutes, completely remove protein from aqueous solutions of single- and double-stranded nucleic acids.
RapidClean is a novel affinity resin designed to remove all protein from aqueous solutions of nucleic acids, providing an alternative to hazardous and lengthy phenol-chloroform procedures. The RapidClean resin binds and removes protein in a 5-minute protocol that combines convenience, speed, and nucleic acid recovery rates in excess of 95%.
Ordering information: Kit includes RapidClean resin and spin filters.
4 Options Available Below
Product Details & Documents
Rapid clean is the fast and easy way to remove protein from RNA and DNA samples.
- Complete removal of proteins from aqueous solutions of nucleic acids
- Protocol takes less than 5 min
- Provides non-toxic alternative to phenol-chloroform extraction
- Nucleic acid recovery rates >95%
- Recovers nucleic acids from 5 to greater than 40 kb
- Suitable for DNA, RNA, cDNA, microRNA, and oligonucleotides
In less than 5 minutes, completely remove protein from aqueous solutions of single- and double-stranded nucleic acids.
RapidClean is a novel affinity resin designed to remove all protein from aqueous solutions of nucleic acids, providing an alternative to hazardous and lengthy phenol-chloroform procedures. The RapidClean resin binds and removes protein in a 5-minute protocol that combines convenience, speed, and nucleic acid recovery rates in excess of 95%.
Ordering information: Kit includes RapidClean resin and spin filters.
Documents
Product Family Options
Product Information
- SizeAvailabilityPrice
- 103254-754RapidClean Protein Removal KitSupplier Number: K-01001-010Specifications:MarketsourceMarketsource
- 103254-756RapidClean Protein Removal KitSupplier Number: K-01001-025Specifications:MarketsourceMarketsource
- 103254-758RapidClean ResinSupplier Number: R-14011-250Specifications:MarketsourceMarketsource
- 103254-760RapidClean ResinSupplier Number: R-14011-B10Specifications:MarketsourceMarketsource