About this item
Nitrocellulose and PVDF membranes for Western blotting.
- High performance: quality controlled for background-free Western blots
- Conveniently available as pre-cut or rolls
- High signal-to-noise ratio for clearer results
- Flexibility to choose the membrane right for your applications
Advansta offers a large selection of nitrocellulose and polyvinylidene difluoride (PVDF) transfer membranes for immunoblotting applications
The WesternBright PVDF membranes provide durability and high tensile strength for ease of handling. Advansta offers PVDF membranes appropriate for chemiluminescent detection (PVDF-CL) and low-autofluorescence PVDF membranes (PVDF-FL) perfect for fluorescent detection.
The WesternBright nitrocellulose membranes are pure nitrocellulose media that has high binding capacity for proteins. These nitrocellulose membranes are stronger and more durable than other nitrocellulose membranes, and will not rip, tear, or crack during transfers. The nature of this material yields sharper images with little or no distortion.
Choose from convenient pre-cut membranes sized for protein minigels and economical membrane rolls. All membranes are optimized for compatibility with a wide range of protein stains and enzyme-based detection systems, specifically with chemiluminescent substrates.