ContactUsLinkComponent
Fluorescent Western Blot for Standardization, Advansta
Catalog # 103254-718
Supplier:  ADVANSTA INC MS
Fluorescent Western Blot for Standardization, Advansta
Catalog # 103254-718
Supplier:  ADVANSTA INC MS
Supplier Number:  K-08001-001
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Fluorescent Western Standardization Blot
  • Cat. No.:
    103254-718
  • Supplier no.:
    K-08001-001
Specifications
About this item
Three-color fluorescent blot ready to image with red/green/blue channels.
  • Fluorescently labeled primary and secondary antibodies on the provided membrane are detectable in red, green, and blue fluorescent channels (equivalent to Cy2/3/5)
  • Convenient for evaluating imager performance
  • Reliable and quality controlled with lot-specific statistics provided
This pre-processed Western blot was developed as a tool to verify the performance of fluorescence imaging systems, including laser scanners and CCD-based instruments
Gel processing: Proteins were separated by polyacrylamide gel electrophoresis followed by transfer to a low-fluorescent PVDF membrane. Two proteins, tubulin and transferrin were detected by primary antibodies made in rat and rabbit hosts, followed by secondary antibodies labeled with different fluorescent dyes. Next, GAPDH was detected by a goat primary antibody directly labeled with SpectraDye Antibody Labeling Kit-Dye 490. The included background quenching sheet with low auto-fluorescence should be used for blot imaging to greatly reduce general background, especially in the blue channel
Antibodies used on the blot fluoresce in the red, green, and blue fluorescent channels.