ContactUsLinkComponent
Hydrolysis of Salts Student Activity
  470003-676
 :  Lab-Aids
undefined
Hydrolysis of Salts Student Activity
  470003-676
 :  Lab-Aids
 :  112
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Hydrolysis of Salts Student Activity
  • No. of lab groups:
    12
  • Cat. No.:
    470003-676
  • Includes:
    Teacher's Guide, Student Guides and Worksheets, SDS
  • Supplier no.:
    112
Determine the Effect of Hydrolysis in Producing Acids or Bases.
  • Determine relative pH and strength of various salt solutions
  • Requires one 50 minute class period
Students discover how and why salts vary in neutrality as they determine actual acidity or basicity of different salt solutions. With the use of the Chemplates® each student tests seven different salt solutions for pH, predicts relative strength, and determines whether the salt hydrolyzed in water produces an acidic or basic solution. This is all done through observation.
 : Materials Include CHEMPLATES with spatula, Deionized Water, Aluminum Sulfate, Ammonium Chloride, Copper II Nitrate, Potassium Sulfate, Sodium Acetate, Sodium Carbonate, Sodium Chloride, Universal pH indicator paper.